EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
TPP and TCEP induce oxidative stress and alter steroidogenesis in TM3 Leydig cells.
PMID 26049154
Abstract
Effects of triphenyl phosphate (TPP) and tris-(2-chloroethyl) phosphate (TCEP) exposure on induction of oxidative stress and endocrine disruption were investigated in TM3 cells. After 24h exposure, cell growth declined and morphology changed in TPP and TCEP treated groups with high dosages. Significant increases in superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPX) and glutathione S-transferase (GST) activities and their respective gene expressions in a dose-dependent and/or time-dependent manner in TPP or TCEP groups. Moreover, the expression of main genes related to testosterone (T) synthesis including cytochrome P450 cholesterol side-chain cleavage enzyme (P450scc), cytochrome P450 17α-hydroxysteroid dehydrogenase (P450-17α), 3β-hydroxysteroid dehydrogenase (3β-HSD) and 17β-hydroxysteroid dehydrogenase (17β-HSD) were dramatically reduced by TPP and TCEP treatments, especially with the high dosage for 24h. TPP and TCEP treatments for 24h caused significant decreases in T levels in the medium. Furthermore, co-treatments of hCG with TPP or TCEP could inhibit hCG-induced changes in the expression of P450scc, P450-17α and 17β-HSD and T levels. Taken together, TPP and TCEP could induce oxidative stress and endocrine disruption in TM3 cells.