EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Transient and quasi-permanent networks in xyloglucan solutions.
PMID 26050908
Abstract
Viscoelastic properties of aqueous solutions of xyloglucan extracted from Hymenaea courbaril seeds (Jatobá gum) were investigated by rheology over a wide range of concentrations and temperatures. The polymer was characterized in dilute solutions by light scattering measurements and size exclusion chromatography. Xyloglucan formed, in semi-dilute solutions (C 0.3 wt%), a transient network with cross-links characterized by a broad distribution of lifetimes, independent of the temperature and concentration. Progressively, at higher temperatures (>60°C), a second much weaker quasi-permanent network was formed and attributed to the exchange of intra- to inter-chain bonds. The stiffness of the second network increased with decreasing temperature, but it could be easily broken by applying a relatively weak shear stress and is readily reversible on re-heating, and partially reversible on resting at 20°C.