EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
New thiocyanate-free ruthenium(II) sensitizers with different pyrid-2-yl tetrazolate ligands for dye-sensitized solar cells.
PMID 26051002
Abstract
The synthesis of two new thiocyanate free Ru(II) complexes with different pyrid-2-yl tetrazolate ligands is reported, along with their electrochemical, spectroscopic and theoretical characterization. The corresponding dye-sensitized solar cell devices were prepared, leading to 3.4% conversion efficiency, promising data, considering the simplicity of the ligands and the high chemical stability of the complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281026
Tetrabutylammonium hexafluorophosphate, 98%
C16H36F6NP