EMAIL THIS PAGE TO A FRIEND
Experimental gerontology
Lifelong wheel running exercise and mild caloric restriction attenuate nuclear EndoG in the aging plantaris muscle.
PMID 26055450
Abstract
Apoptosis plays an important role in atrophy and sarcopenia in skeletal muscle. Recent evidence suggests that insufficient heat shock proteins (HSPs) may contribute to apoptosis and muscle wasting. In addition, long-term caloric restriction (CR) and lifelong wheel running exercise (WR) with CR provide significant protection against caspase-dependent apoptosis and sarcopenia. Caspase-independent mediators (endonuclease G: EndoG; apoptosis-inducing factor: AIF) of apoptosis are also linked to muscles wasting with disuse and aging. However, the efficacy of CR and WR with CR to attenuate caspase-independent apoptosis and preserve HSPs in aging skeletal muscle are unknown. Therefore, we tested the hypothesis that CR and WR with CR would ameliorate age-induced elevation of EndoG and AIF while protecting HSP27 and HSP70 levels in the plantaris. Male Fischer-344 rats were divided into 4 groups at 11weeks: ad libitum feeding until 6months (YAL); fed ad libitum until 24months old (OAL); 8%CR to 24months (OCR); WR+8%CR to 24months (OExCR). Nuclear EndoG levels were significantly higher in OAL (+153%) than in YAL, while CR (-38%) and WR with CR (-46%) significantly attenuated age-induced increment in nuclear EndoG. HSP27 (-63%) protein content and phosphorylation at Ser82 (-49%) were significantly lower in OAL than in YAL, while HSP27 protein content was significantly higher in OCR (+136%) and OExCR (+155%) and p-HSP27 (+254%) was significantly higher in OExCR compared with OAL, respectively. In contrast, AIF and HSP70 were unaltered by CR or WR with CR in aging muscle. These data indicate that CR and WR with CR attenuate age-associated upregulation of EndoG translocation in the nucleus, potentially involved with HSP27 signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS2301
Anti-AIF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500441
Anti-AIF antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406783
Anti-AIFM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100079
Anti-AIFM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA030611
Anti-AIFM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021335
Anti-ENDOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021830
Anti-ENDOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2103518
Anti-ENDOG antibody produced in rabbit, affinity isolated antibody
SAB1405757
Anti-ENDOG, (C-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
E5654
Anti-Endonuclease G antibody produced in rabbit, 1 mg/mL, affinity isolated antibody, buffered aqueous solution
H4163
Anti-Heat Shock Factor 1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300509
Anti-HSF1 (Ab-303) antibody produced in rabbit, affinity isolated antibody
SAB4300466
Anti-HSF1 (Ab-307) antibody produced in rabbit, affinity isolated antibody
AV100769
Anti-HSF1 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV36894
Anti-HSF1 (AB2) antibody produced in rabbit, affinity isolated antibody
HPA008888
Anti-HSF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36895
Anti-HSF1 antibody produced in rabbit, IgG fraction of antiserum
SAB4501448
Anti-HSF1 antibody produced in rabbit, affinity isolated antibody
SAB4501449
Anti-HSF1 antibody produced in rabbit, affinity isolated antibody
SAB4501450
Anti-HSF1 antibody produced in rabbit, affinity isolated antibody
SAB1306362
ANTI-HSP27 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501456
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4501457
Anti-HSP27 antibody produced in rabbit, affinity isolated antibody
SAB4300495
Anti-HSPB1 (Ab-78) antibody produced in rabbit, affinity isolated antibody
SAB4300496
Anti-HSPB1 (Ab-82) antibody produced in rabbit, affinity isolated antibody
AV09060
Anti-HSPB1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV30177
Anti-HSPB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA000497
Anti-HSPB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701965
Anti-HSPB1 antibody produced in rabbit
HPA038889
Anti-HSPB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution