EMAIL THIS PAGE TO A FRIEND
Journal of Zhejiang University. Science. B
Heat stress inhibits proliferation, promotes growth, and induces apoptosis in cultured Lantang swine skeletal muscle satellite cells.
PMID 26055917
Abstract
Proliferation suppression and apoptosis are the prominent characteristics induced by heat stress (HS) in cells, whereas the effects of HS on cell growth (mass accumulation) are unknown. In this study, Lantang swine (an indigenous breed of China) skeletal muscle satellite cells (SCs) were pre-cultured at 37 °C for 24 h. The HS group was subjected to HS at 41 °C, while the control group was maintained at 37 °C. Heat shock protein 70 (HSP70) expression and SC size are significantly increased (P<0.05) by HS, but cell proliferation is suppressed (P<0.05) and apoptosis is induced (P<0.05). HS led to a lower percentage of SCs in the G0/G1 phase (P<0.05) together with a higher percentage of SCs in the S phase (P<0.05). However, the percentage of SCs in the G2/M phase was decreased (P<0.05) at 48 h but then increased (P<0.05) at 72 h with HS. In addition, the phosphorylation ratios of protein kinase b (Akt), ribosomal protein S6 kinase (S6K), and ribosomal protein S6 were increased (P<0.05) by HS. Nevertheless, the phosphorylation ratios of the 4E binding protein 1 and the eukaryotic initiation factor-4E were indistinguishable (P>0.05) from those of the control group. The phosphorylation ratio of the mammalian target of rapamycin (mTOR) (Ser(2448)) increased (P<0.05) within 48 h, and apparent differences were abrogated at 72 h (P>0.05). Moreover, cleaved caspase-3 expression was increased at 72 h (P<0.05). These findings indicate that HS induces apoptosis and disrupts cell cycle distribution to decrease the number of cells. Additionally, HS can promote SC growth via an activated Akt/mTOR/S6K signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
SAB4503847
Anti-phospho-4E-BP1 (pSer64) antibody produced in rabbit, affinity isolated antibody
SAB4504328
Anti-phospho-4E-BP1 (pThr36) antibody produced in rabbit, affinity isolated antibody
SAB4504329
Anti-phospho-4E-BP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4503848
Anti-phospho-4E-BP1 (pThr69) antibody produced in rabbit, affinity isolated antibody
SAB4504686
Anti-phospho-4E-BP1 (pThr70) antibody produced in rabbit, affinity isolated antibody
SAB4300179
Anti-phospho-EIF4EBP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4300214
Anti-phospho-RPS6KB1 (pSer411) antibody produced in rabbit, affinity isolated antibody
SAB4300225
Anti-phospho-RPS6KB1 (pSer424) antibody produced in rabbit, affinity isolated antibody
SAB4300202
Anti-phospho-RPS6KB1 (pThr421) antibody produced in rabbit, affinity isolated antibody
SAB4300444
Anti-RPS6KB1 (Ab-389) antibody produced in rabbit, affinity isolated antibody
SAB4300515
Anti-RPS6KB1 (Ab-411) antibody produced in rabbit, affinity isolated antibody
SAB4300500
Anti-RPS6KB1 (Ab-421) antibody produced in rabbit, affinity isolated antibody
SAB4300527
Anti-RPS6KB1 (Ab-424) antibody produced in rabbit, affinity isolated antibody
SAB1412617
ANTI-RPS6KB1 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1412626
ANTI-RPS6KB1 antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
SAB2104784
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2104785
Anti-RPS6KB1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0001978M1
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 4F3-H2, purified immunoglobulin, buffered aqueous solution
SAB1403761
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution
SAB5300484
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 9E12D9, ascites fluid
SAB5300486
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 11G12C11, ascites fluid
SAB1404335
Monoclonal Anti-RPS6KB1, (C-terminal) antibody produced in mouse, clone 1E10, purified immunoglobulin, buffered aqueous solution