EMAIL THIS PAGE TO A FRIEND
British journal of cancer
miR-30e* is an independent subtype-specific prognostic marker in breast cancer.
PMID 26057454
Abstract
Breast cancer clinical outcome is affected by tumor molecular features, and the identification of subtype-specific prognostic biomarkers is relevant for breast cancer translational research. Gene expression signatures proved to be able to complement prognostic information provided by classical clinico-pathological features. Recently, microRNAs (miRNAs) have been causally linked to tumorigenesis and cancer progression and have been associated with patient outcome, also in breast cancer. MicroRNAs associated with the development of distant metastasis were identified in a cohort of 92 ESR1+/ERBB2- lymph node-negative breast cancers from patients not receiving adjuvant treatment. Results were confirmed and further investigated in a total of 1246 miRNA and gene expression profiles of the Molecular Taxonomy of Breast Cancer International Consortium data set. Moderated t-test, univariable and multivariable Cox regression models were used for statistical analyses. miR-30e* was identified as independent protective prognostic factor in lymph node-negative untreated patients with ESR1+/ERBB2- tumours and retained a significant association with a good prognosis in treated patients with the same tumor subtype as well as in the ERBB2+ subtype, but not in ESR1-/ERBB2- tumours. We highlighted a relevant and subtype-specific role in breast cancer for miR-30e* and demonstrated that adding miRNA markers to gene signatures and clinico-pathological features can help for a better prognostication.