EMAIL THIS PAGE TO A FRIEND
Oncotarget
Quercetin-induced apoptosis prevents EBV infection.
PMID 26059439
Abstract
Epstein-Barr virus (EBV) is a human gamma-1 herpesvirus that establishes a lifelong latency in over 90% of the world's population. During latency, virus exists predominantly as a chromatin-associated, multicopy episome in the nuclei of a variety of tumor cells derived from B cells, T cells, natural killer (NK) cells, and epithelial cells. Licorice is the root of Glycyrrhiza uralensis or G. glabra that has traditionally cultivated in eastern part of Asia. Licorice was reported to have anti-viral, anti-inflammatory, anti-atopic, hepatoprotective, anti-neurodegenerative, anti-tumor, anti-diabetic effects and so forth. Quercetin and isoliquiritigenin are produced from licorice and highly similar in molecular structure. They have diverse bioactive effects such as antiviral activity, anti-asthmatic activity, anti-cancer activity, anti-inflammation activity, monoamine-oxidase inhibitor, and etc. To determine anti-EBV and anti-EBVaGC (Epstein-Barr virus associated gastric carcinoma) effects of licorice, we investigated antitumor and antiviral effects of quercetin and isoliquiritigenin against EBVaGC. Although both quercetin and isoliquiritigenin are cytotoxic to SNU719 cells, quercetin induced more apoptosis in SNU719 cells than isoliquiritigenin, more completely eliminated DNMT1 and DNMT3A expressions than isoliquiritigenin, and more strongly affects the cell cycle progression of SNU719 than isoliquiritigenin. Both quercetin and isoliquiritigenin induce signal transductions to stimulate apoptosis, and induce EBV gene transcription. Quercetin enhances frequency of F promoter use, whereas isoliquiritigenin enhances frequency of Q promoter use. Quercetin reduces EBV latency, whereas isoliquiritigenin increases the latency. Quercetin increases more the EBV progeny production, and inhibits more EBV infection than isoliquiritigenin. These results indicate that quercetin could be a promising candidate for antiviral and antitumor agents against EBV and human gastric carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS3805
Anti-AIM (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3807
Anti-AIM (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22659 Anti-DNMT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500321
Anti-DNMT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
D4692
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D4567
Anti-DNMT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA002694 Anti-DNMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106406
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody
D8695
Anti-DNMT3A (611-625) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501730
Anti-DNMT3A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405725
Anti-DNMT3A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA026588
Anti-DNMT3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410305
Anti-DNMT3A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22661 Anti-DNMT3A2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300497
Anti-MAPK9 (Ab-183) antibody produced in rabbit, affinity isolated antibody
GW22791 Anti-MAPK9 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2502040
Anti-MAPK9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502041
Anti-MAPK9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412229
ANTI-MAPK9 antibody produced in mouse, clone 3C12, purified immunoglobulin, buffered aqueous solution
SAB4502402
Anti-MAPK9 antibody produced in rabbit, affinity isolated antibody
SAB2107297
Anti-MAPK9 antibody produced in rabbit, affinity isolated antibody
SAB4300198
Anti-phospho-MAPK9 (pThr183) antibody produced in rabbit, affinity isolated antibody
SAB4502399
Anti-SAPK antibody produced in rabbit, affinity isolated antibody
SAB4502400
Anti-SAPK antibody produced in rabbit, affinity isolated antibody
I3766
Isoliquiritigenin, powder
C15H12O4
SAB1403742
Monoclonal Anti-DNMT1, (N-terminal) antibody produced in mouse, clone 2B5, ascites fluid
SAB1404250
Monoclonal Anti-MAPK9, (C-terminal) antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
PLA0011
Rabbit anti-DNMT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.