EMAIL THIS PAGE TO A FRIEND
Nutrients
Quercetin Decreases Claudin-2 Expression Mediated by Up-Regulation of microRNA miR-16 in Lung Adenocarcinoma A549 Cells.
PMID 26061016
Abstract
Claudin-2 is highly expressed in human lung adenocarcinoma tissues and cells. Knockdown of claudin-2 decreases cell proliferation and migration. Claudin-2 may be a novel target for lung adenocarcinoma. However, there are no physiologically active substances of foods which decrease claudin-2 expression. We here found that quercetin, a flavonoid present in fruits and vegetables, time- and concentration-dependently decreases claudin-2 expression in lung adenocarcinoma A549 cells. In the present study, we examined what regulatory mechanism is involved in the decrease in claudin-2 expression by quercetin. Claudin-2 expression was decreased by LY-294002, a phosphatidylinositol 3-kinase (PI3-K) inhibitor, and U0126, a MEK inhibitor. These drugs inhibited the phosphorylation of Akt and ERK1/2, which are downstream targets of PI3-K and MEK, respectively. In contrast, quercetin did not inhibit the phosphorylation. Both LY-294002 and U0126 inhibited promoter activity of claudin-2, but quercetin did not. The stability of claudin-2 mRNA was decreased by quercetin. Quercetin increased the expression of microRNA miR-16. An inhibitor of miR-16 rescued quercetin-induced decrease in the claudin-2 expression. These results suggest that quercetin decreases claudin-2 expression mediated by up-regulation of miR-16 expression and instability of claudin-2 mRNA in lung adenocarcinoma cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503545
Anti-Claudin 1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503546
Anti-Claudin 1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503544
Anti-Claudin 2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500436
Anti-Claudin 7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500437
Anti-Claudin 7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4200534
Anti-Claudin-1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200462
Anti-Claudin-1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1406776
Anti-CLDN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV33623
Anti-CLDN1 antibody produced in rabbit, affinity isolated antibody
SAB3500438
Anti-CLDN1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV42845
Anti-CLDN2 antibody produced in rabbit, IgG fraction of antiserum
SAB2100443
Anti-CLDN7 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100444
Anti-CLDN7 (ab2) antibody produced in rabbit, affinity isolated antibody
HPA014703
Anti-CLDN7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C3394
Cordycepin, from Cordyceps militaris
C10H13N5O3
WH0009076M1
Monoclonal Anti-CLDN1 antibody produced in mouse, clone 1C5-D9, purified immunoglobulin, buffered aqueous solution
SAB1404666
Monoclonal Anti-CLDN1 antibody produced in mouse, clone 2E2-H5, purified immunoglobulin, buffered aqueous solution
WH0009075M1
Monoclonal Anti-CLDN2 antibody produced in mouse, clone 3F1, purified immunoglobulin, buffered aqueous solution