PloS one
IFN-Gamma Inhibits JC Virus Replication in Glial Cells by Suppressing T-Antigen Expression.
PMID 26061652
Abstract
Patients undergoing immune modulatory therapies for the treatment of autoimmune diseases such as multiple sclerosis, and individuals with an impaired-immune system, most notably AIDS patients, are in the high risk group of developing progressive multifocal leukoencephalopathy (PML), an often lethal disease of the brain characterized by lytic infection of oligodendrocytes in the central nervous system (CNS) with JC virus (JCV). The immune system plays an important regulatory role in controlling JCV reactivation from latent sites by limiting viral gene expression and replication. However, little is known regarding the molecular mechanisms responsible for this regulation. Here, we investigated the impact of soluble immune mediators secreted by activated PBMCs on viral replication and gene expression by cell culture models and molecular virology techniques. Our data revealed that viral gene expression and viral replication were suppressed by soluble immune mediators. Further studies demonstrated that soluble immune mediators secreted by activated PBMCs inhibit viral replication induced by T-antigen, the major viral regulatory protein, by suppressing its expression in glial cells. This unexpected suppression of T-antigen was mainly associated with the suppression of translational initiation. Cytokine/chemokine array studies using conditioned media from activated PBMCs revealed several candidate cytokines with possible roles in this regulation. Among them, only IFN-γ showed a robust inhibition of T-antigen expression. While potential roles for IFN-β, and to a lesser extent IFN-α have been described for JCV, IFN-γ has not been previously implicated. Further analysis of IFN-γ signaling pathway revealed a novel role of Jak1 signaling in control of viral T-antigen expression. Furthermore, IFN-γ suppressed JCV replication and viral propagation in primary human fetal glial cells, and showed a strong anti-JCV activity. Our results suggest a novel role for IFN-γ in the regulation of JCV gene expression via downregulation of the major viral regulatory protein, T-antigen, and provide a new avenue of research to understand molecular mechanisms for downregulation of viral reactivation that may lead to development of novel strategies for the treatment of PML.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407036
Anti-KHDRBS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV40665
Anti-KHDRBS1 antibody produced in rabbit, affinity isolated antibody
AV37130
Anti-KHDRBS1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104552
Anti-KHDRBS1 antibody produced in rabbit, affinity isolated antibody
SAB4503847
Anti-phospho-4E-BP1 (pSer64) antibody produced in rabbit, affinity isolated antibody
SAB4504328
Anti-phospho-4E-BP1 (pThr36) antibody produced in rabbit, affinity isolated antibody
SAB4504329
Anti-phospho-4E-BP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4503848
Anti-phospho-4E-BP1 (pThr69) antibody produced in rabbit, affinity isolated antibody
SAB4504686
Anti-phospho-4E-BP1 (pThr70) antibody produced in rabbit, affinity isolated antibody
SAB4300179
Anti-phospho-EIF4EBP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
S9575
Anti-Sam68 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
S9450
Anti-Sam68 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406748
Anti-SQSTM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3500430
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104334
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody
SAB3500594
Anti-ZIP3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0010657M3
Monoclonal Anti-KHDRBS1 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution
WH0008878M1
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
SAB1404647
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution