EMAIL THIS PAGE TO A FRIEND
Oncology reports
Butorphanol, a synthetic opioid, sensitizes ABCB1-mediated multidrug resistance via inhibition of the efflux function of ABCB1 in leukemia cells.
PMID 26062728
Abstract
Multidrug resistance (MDR) remains a formidable challenge in the use of chemotherapy and represents a powerful obstacle to the treatment of leukemia. ATP-binding cassette subfamily B member 1 (ABCB1) is a recognized factor which causes MDR and is closely related to poor outcome and relapse in leukemia. Ongoing research concerning the strategy for inhibiting the abnormally high activity of the ABCB1 transporter is critically needed. In the present study, we sought to elucidate the interaction between ABCB1 transporter and butorphanol. Our results showed that butorphanol significantly antagonized ABCB1-mediated drug efflux and increased the intracellular drug concentration by inhibiting the transport activity of ABCB1 in leukemia cells. Mechanistic investigations demonstrated that butorphanol did not alter the protein expression or localization of ABCB1 in HL60/VCR and K562/ADR cells. Furthermore, homology modeling indicated that butorphanol could fit into the large drug-binding cavity of ABCB1 and form a binding conformation. In conclusion, butorphanol reversed the ABCB1-mediated MDR in leukemia cells by directly suppressing the efflux activity of ABCB1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V8388
Vincristine sulfate, meets USP testing specifications
C46H56N4O10 · H2SO4
V8879
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder
C46H56N4O10 · H2SO4