EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
Ginsenoside Rg1 reduces aldosterone-induced autophagy via the AMPK/mTOR pathway in NRK-52E cells.
PMID 26063203
Abstract
Aldosterone is a steroid hormone secreted from the adrenal cortex, which regulates blood pressure. Higher concentrations of aldosterone can cause several diseases, including hypertension, diabetic nephropathy and chronic kidney disease. Previous reports have demonstrated that aldosterone has a pathogenic role in renal injury via reactive oxygen species (ROS), which involves the regulation of autophagy. However, whether aldosterone can induce autophagy in renal tubular cells remains to be elucidated. In the present study, elevated autophagy was observed in rat renal tubular NRK-52E cells exposed to aldosterone, which was demonstrated by the increased number of autophagosomes, conversion of LC3-I to LC3-II and the expression of Beclin-1. The enhanced autophagy was accompanied by increased production of intracellular ROS, which was reversed by N-acetylcysteine, a specific inhibitor of ROS signaling. Furthermore, treatment with ginsenoside Rg1 reduced the aldosterone-induced autophagy and production of ROS, possibly through reducing the phosphorylation of AMPK and preserving mTOR activity. These findings demonstrated that aldosterone promoted ROS generation and increased autophagy in the NRK-52E cells. Ginsenoside Rg1 effectively relieved aldosterone-induced oxidative stress and abnormal autophagy, suggesting that Rg1 may be used as a potential therapeutic drug to inhibit the renal injury, which is induced by aldosterone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700864 Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700865 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700862 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700860 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700866 Anti-Rabbit IgG (Fc specific)-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700979 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700977 Anti-Rabbit IgG (H+L), F(ab) fragment-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700975 Anti-Rabbit IgG (H+L), F(ab) fragment-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700980 Anti-Rabbit IgG (H+L), F(ab) fragment-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB4600003 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600010
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 350 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600036
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600061
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 555 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600076
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 568 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600099
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 594 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600132
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 633 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600155
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 640R antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600177
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 647 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600208
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 680R antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600372 Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF750 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600373 Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF750 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3700935 Anti-Rabbit IgG (H+L)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700936 Anti-Rabbit IgG (H+L)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700933 Anti-Rabbit IgG (H+L)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700925 Anti-Rabbit IgG (H+L)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700937
Anti-Rabbit IgG (H+L)-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder