EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Direct Chemical Activation of a Rationally Engineered Signaling Enzyme.
PMID 26063205
Abstract
Few chemical strategies for activating enzymes have been developed. Here we show that a biarsenical compound (FlAsH) can directly activate a rationally engineered protein tyrosine phosphatase (Shp2 PTP) by disrupting autoinhibitory interactions between Shp2's N-terminal SH2 domain and its PTP domain. We found that introducing a tricysteine motif at a loop of Shp2's N-SH2 domain confers affinity for FlAsH; binding of FlAsH to the cysteine-enriched loop relieves Shp2's inhibitory interdomain interaction and substantially increases the enzyme's PTP activity. Activation of engineered Shp2 is substrate independent and is observed in the contexts of both purified enzyme and complex proteomes. A chemical means for activating Shp2 could be useful for investigating its roles in signaling and oncogenesis, and the loop-targeting strategy described herein could provide a blueprint for the development of target-specific activators of other autoinhibited enzymes.