EMAIL THIS PAGE TO A FRIEND
Molecular neurobiology
Melatonin Ameliorates Arsenite-Induced Neurotoxicity: Involvement of Autophagy and Mitochondria.
PMID 26063592
Abstract
In the present study, the neuroprotective effect of melatonin on arsenite-induced neurotoxicity was investigated in rat primary cultured cortical neurons. Incubation of melatonin prevented arsenite-induced neuronal cell loss in a concentration-dependent manner. Furthermore, melatonin significantly attenuated arsenite-induced elevation in microtubule-associated protein light chain 3 (LC3)-II levels, a biomarker of autophagy. Our fluorescent staining assay showed that melatonin decreased arsenite-induced elevation of co-localized fluorescent puncta of monodansylcadaverine (a specific marker of autophagic vacuoles) and lysotracker red (a specific marker of lysosomes), indicating that melatonin is capable of inhibiting arsenite-induced autophagy and autolysosome formation. Because 3-methyladenine (an autophagic inhibitor) attenuated the arsenite-reduced α-synuclein levels (a protein essential for the neurite outgrowth and synaptic plasticity), melatonin via inhibiting autophagy attenuated the arsenite-reduced α-synuclein levels. At the same time, melatonin ameliorated the arsenite-induced reduction in growth associated protein 43 (a hallmark protein of neurite outgrowth) and discontinuous neurites of rat primary cultured cortical neurons. In addition, melatonin was found to prevent arsenite-induced decreases in cytochrome c oxidase levels (a biomarker of mitochondrial mass) and elevation in co-localized fluorescent puncta of autolysosomes and cytochrome c oxidase. Moreover, melatonin prevented arsenite-induced reduction in peroxisome proliferator-activated receptor gamma co-activator 1 α, a transcriptional co-activator of mitochondrial biosynthesis. Taken together, melatonin may exert its neuroprotective action via inhibiting arsenite-induced autophagy and enhancing mitochondrial biogenesis and thus restoring α-synuclein levels, neuronal integrity, and mitochondrial mass in rat primary cultured cortical neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100769
Anti-FAM82A (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100770
Anti-FAM82A (ab2) antibody produced in rabbit, affinity isolated antibody
SAB3500133
Anti-FAM82A1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2103283
Anti-FAM82B, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1407628
Anti-FAM82C antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2103199
Anti-FAM82C antibody produced in rabbit, affinity isolated antibody
SAB1305768
ANTI-RMD2 (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA026495
Anti-RMDN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA034706
Anti-RMDN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009975
Anti-RMDN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA022008
Anti-SPAG5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA022479
Anti-SPAG5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA053057
Anti-SYBU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0010615M1
Monoclonal Anti-SPAG5 antibody produced in mouse, clone 5F9, purified immunoglobulin, buffered aqueous solution
SAB1404850
Monoclonal Anti-SPAG5 antibody produced in mouse, clone 5F9, ascites fluid