EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental medicine
Effects of Foxp3 gene modified dendritic cells on mouse corneal allograft rejection.
PMID 26064298
Abstract
To investigate the effect of Foxp3 gene modified dendritic cells (Foxp3 + DC) on allogeneic T cells proliferation and to study the effect of Foxp3 + DC on corneal allograft rejection. Lentivirus-Foxp3 was transfected into DC2.4 cells, as Foxp3 + DC cells. 42 BALB/c mice were randomly divided into: Group A (n = 6), normal group; Group B (n = 12), Group C (n = 12) and Group D (n = 12), allograft groups, were treated with normal saline, DC2.4, Foxp3 + DC by intraperitoneal injection, respectively. Compared with the control group, Foxp3 protein in the Foxp3 + DC cells increased significantly (P < 0.05); the expressions of CD80 and CD86 immunophenotypes of Foxp3 + DC cells decreased significantly (P < 0.05); IL-12 secretion reduced (P < 0.05), but IL-10 secretion was promoted (P < 0.05). The average transplant survival time in Group B was (14.833 ± 1.472) d, and Group C and Group D led to a statistically significant prolongation of transplant survival to (17.667 ± 1.366, 23.000 ± 2.000) d (P < 0.05) respectively. 14 d after transplantation, as compared with Group C and D, the expressions of IFN-γ in grafts markedly increased in Group B. 14 d after transplantation, as compared with Group B, the expressions of Foxp3 mRNA, IDO mRNA in grafts decreased remarkably in Group C and D (P < 0.05); as compared with Group C, the expressions of Foxp3 mRNA, IDO mRNA in grafts decreased remarkably in Group D (P < 0.05). Foxp3 + DC cells reduce the expression of costimulatory factors, reduce the secretion of IL-12, promote IL-10 production and inhibit the stimulation of alloreactive T cell proliferation response capacity. Foxp3 + DC cells play important roles in inhibiting corneal allograft immune response and prolonging graft survival time.