EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Production of prostaglandin E2 induced by cigarette smoke modulates tissue factor expression and activity in endothelial cells.
PMID 26065856
Abstract
Cigarette smoke (CS) increases the incidence of atherothrombosis, the release of prostaglandin (PG) E2, and the amount of tissue factor (TF). The link between PGE2 and TF, and the impact of this interaction on CS-induced thrombosis, is unknown. Plasma from active smokers showed higher concentration of PGE2, TF total antigen, and microparticle-associated TF (MP-TF) activity compared with never smokers. Similar results were obtained in mice and in mouse cardiac endothelial cells (MCECs) after treatment with aqueous CS extracts (CSEs) plus IL-1β [CSE (6.4 puffs/L)/IL-1β (2 μg/L)]. A significant correlation between PGE2 and TF total antigen or MP-TF activity were observed in both human and mouse plasma or tissue. Inhibition of PGE synthase reduced TF in vivo and in vitro and prevented the arterial thrombosis induced by CSE/IL-1β. Only PG E receptor 1 (EP1) receptor antagonists (SC51089:IC50 ∼ 1 μM, AH6809:IC50 ∼ 7.5 μM) restored the normal TF and sirtuin 1 (SIRT1) levels in MCECs before PGE2 (EC50 ∼ 2.5 mM) or CSE/IL-1β exposure. Similarly, SIRT1 activators (CAY10591: IC50 ∼ 10 μM, resveratrol: IC50 ∼ 5 μM) or prostacyclin analogs (IC50 ∼ 5 μM) prevented SIRT1 inhibition and reduced TF induced by CSE/IL-1β or by PGE2. In conclusion, PGE2 increases both TF expression and activity through the regulation of the EP1/SIRT1 pathway. These findings suggest that EP1 may represent a possible target to prevent prothrombotic states.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6807
Misoprostol, ≥99% (HPLC)
C22H38O5
S7942
Sirtinol, ≥95% (NMR)
C26H22N2O2