EMAIL THIS PAGE TO A FRIEND
Nature communications
Loss of microRNA-27b contributes to breast cancer stem cell generation by activating ENPP1.
PMID 26065921
Abstract
Cancer stem cells (CSCs) have been identified in various types of cancer; however, the mechanisms by which cells acquire CSC properties such as drug resistance and tumour seeding ability are not fully understood. Here, we identified microRNA-27b (miR-27b) as a key regulator for the generation of a side-population in breast cancer cells that showed CSC properties, and also found that the anti-type II diabetes (T2D) drug metformin reduced this side-population via miR-27b-mediated repression of ectonucleotide pyrophosphatase/phosphodiesterase family member 1 (ENPP1), which is involved in T2D development. ENPP1 induced the generation of the side-population via upregulation of the ABCG2 transporter. ENPP1 was also identified as a substrate of the 26S proteasome, the activity of which is downregulated in CSCs. Overall, these results demonstrate that a T2D-associated gene plays an important role in tumour development and that its expression is strictly controlled at the mRNA and protein levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D150959
1,1-Dimethylbiguanide hydrochloride, 97%
C4H11N5 · HCl
SAB1406822
Anti-ABCG2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300689
Anti-ABCG2 antibody produced in rabbit, affinity isolated antibody
B7185
Anti-Breast Cancer Resistance Protein antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400199
Anti-ENPP1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2700831
Anti-ENPP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW21421 Anti-MITF antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA003259
Anti-MITF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37977
Anti-MITF antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2101480
Anti-MITF antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2105416
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB4501879
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB1403023
Monoclonal Anti-ABCG2 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
B7684 Monoclonal Anti-Breast Cancer Resistance Protein antibody produced in mouse, 250 μg/mL, clone BXP-34, tissue culture supernatant, aqueous solution
B7059 Monoclonal Anti-Breast Cancer Resistance Protein antibody produced in mouse, ~250 μg/mL, clone BXP-21, tissue culture supernatant, aqueous solution
SAB1402713
Monoclonal Anti-CD24 antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
SAB4700623 Monoclonal Anti-CD24 antibody produced in mouse, clone SN3, purified immunoglobulin, buffered aqueous solution