EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Magnetic studies of mesoporous nanostructured iron oxide materials synthesized by one-step soft-templating.
PMID 26067028
Abstract
A combined magnetization and (57)Fe spin-echo nuclear magnetic resonance (NMR) study has been carried out on mesoporous nanostructured materials consisting of the magnetite (Fe3O4) and maghemite (γ-Fe2O3) phases. Two series of samples were synthesized using a recently developed one-step soft-templating approach with systematic variations in calcination temperature and reaction atmosphere. Nuclear magnetic resonance has been shown to be a valuable tool for distinguishing between the two magnetic iron oxide spinel phases, Fe3O4 and γ-Fe2O3, on the nanoscale as well as monitoring phase transformation resulting from oxidation. For the Fe3O4 and γ-Fe2O3 phases, peaks in the NMR spectra are attributed to Fe in the tetrahedral (A) sites and octahedral (B) sites. The magnetic field dependence of the peaks was observed and confirmed the site assignments. Fe3O4 on a nanoscale readily oxidizes to form γ-Fe2O3 and this was clearly evident in the NMR spectra. As evidenced by transmission electron microscope (TEM) images, the porous mesostructure for the iron oxide materials is formed by a random close-packed aggregation of nanoparticles; correspondingly, superparamagnetic behavior was observed in the magnetic measurements. Although X-ray diffraction (XRD) shows the spinel structure for the Fe3O4 and γ-Fe2O3 phases, unlike NMR, it is difficult to distinguish between the two phases with XRD. Nitrogen sorption isotherms characterize the mesoporous structures of the materials, and yield BET surface area values and limited BJH pore size distribution curves.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
700312
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700320
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700304
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
725331
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725358
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725366
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
747327
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747424
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm diameter, biotin functionalized, 1 mg/mL Fe, dispersion in H2O
Fe3O4
747254
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Fe3O4
747343
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747319
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747300
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747408
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), PEG functionalized, dispersion, 1 mg/mL Fe in H2O
Fe3O4
747416
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), biotin functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
790508
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
518158
Iron(II,III) oxide, 99.99% trace metals basis
Fe3O4
637106
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Fe3O4
310069
Iron(II,III) oxide, powder, <5 μm, 95%
Fe3O4