PloS one
Cardiac Fibrosis Alleviated by Exercise Training Is AMPK-Dependent.
PMID 26068068
Abstract
Regular exercise can protect the heart against external stimuli, but the mechanism is not well understood. We determined the role of adenosine monophosphate-activated protein kinase (AMPK) in regulating swimming exercise-mediated cardiac protection against β-adrenergic receptor overstimulation with isoproterenol (ISO) in mice. Ten-week-old AMPKα2+/+ and AMPKα2-knockout (AMPKα2-/-) littermates were subjected to 4 weeks of swimming training (50 min daily, 6 days a week) or housed under sedentary conditions. The mice received daily subcutaneous injection of ISO (5 mg/kg/d), a nonselective β-adrenergic receptor agonist, during the last 2 weeks of swimming training. Swimming training alleviated ISO-induced cardiac fibrosis in AMPKα2+/+ mice but not AMPKα2-/- mice. Swimming training activated cardiac AMPK in AMPKα2+/+ mice. Furthermore, swimming training attenuated ISO-induced production of reactive oxygen species (ROS) and expression of NADPH oxidase and promoted the expression of antioxidant enzymes in AMPKα2+/+ mice but not AMPKα2-/- mice. In conclusion, swimming training attenuates ISO-induced cardiac fibrosis by inhibiting the NADPH oxidase-ROS pathway mediated by AMPK activation. Our findings provide a new mechanism for the cardioprotective effects of exercise.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405562
Anti-CAT antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2105074
Anti-CAT antibody produced in rabbit, affinity isolated antibody
SAB4503383
Anti-Catalase antibody produced in rabbit, affinity isolated antibody
AV53559
Anti-CRAT antibody produced in rabbit, affinity isolated antibody
HPA020260
Anti-CRAT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA019230
Anti-CRAT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB2501192
Anti-CYBB antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4200118
Anti-CYBB antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
HPA000867
Anti-EIF5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100671
Anti-EIF5 antibody produced in rabbit, affinity isolated antibody
SAB2106431
Anti-EIF5 antibody produced in rabbit, affinity isolated antibody
SAB1410328
Anti-EIF5 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406959
Anti-GLYAT antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100933
Anti-GLYAT antibody produced in rabbit, affinity isolated antibody
SAB1304615 ANTI-NOX4 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA015475
Anti-NOX4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500976
Anti-SOD1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406464
Anti-SOD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001401
Anti-SOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45752
Anti-SOD1 antibody produced in rabbit, IgG fraction of antiserum
SAB5200083
Anti-SOD1 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB1411305
Anti-SOD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501675
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501676
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406465
Anti-SOD2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701618
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701761
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001814
Anti-SOD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102261
Anti-SOD2 antibody produced in rabbit, affinity isolated antibody
HPA062864
Anti-TRPV6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution