EMAIL THIS PAGE TO A FRIEND
Cancer discovery
ARID1A Deficiency Impairs the DNA Damage Checkpoint and Sensitizes Cells to PARP Inhibitors.
PMID 26069190
Abstract
ARID1A, SWI/SNF chromatin remodeling complex subunit, is a recently identified tumor suppressor that is mutated in a broad spectrum of human cancers. Thus, it is of fundamental clinical importance to understand its molecular functions and determine whether ARID1A deficiency can be exploited therapeutically. In this article, we report a key function of ARID1A in regulating the DNA damage checkpoint. ARID1A is recruited to DNA double-strand breaks (DSB) via its interaction with the upstream DNA damage checkpoint kinase ATR. At the molecular level, ARID1A facilitates efficient processing of DSB to single-strand ends and sustains DNA damage signaling. Importantly, ARID1A deficiency sensitizes cancer cells to PARP inhibitors in vitro and in vivo, providing a potential therapeutic strategy for patients with ARID1A-mutant tumors. ARID1A has been identified as one of the most frequently mutated genes across human cancers. Our data suggest that clinical utility of PARP inhibitors might be extended beyond patients with BRCA mutations to a larger group of patients with ARID1A-mutant tumors, which may exhibit therapeutic vulnerability to PARP inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4436
Anti-53BP1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
B4561
Anti-53BP1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2100125
Anti-ANTXR1 antibody produced in rabbit, affinity isolated antibody
HPA052046
Anti-ANTXR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005456
Anti-ARID1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300576
Anti-ATR (Ab-428) antibody produced in rabbit, affinity isolated antibody
PRS3117
Anti-ATR (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3119
Anti-ATR (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200348
Anti-ATR (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA028264
Anti-ATR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700854
Anti-ATRIP (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1409213
Anti-MMAB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA039017
Anti-MMAB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300100
Anti-phospho-ATM (pSer1981) antibody produced in rabbit, affinity isolated antibody
SAB4300116
Anti-phospho-CHEK1 (pSer280) antibody produced in rabbit, affinity isolated antibody
SAB4300121
Anti-phospho-CHEK2 (pSer516) antibody produced in rabbit, affinity isolated antibody
SAB4504363
Anti-phospho-Chk1 (pSer280) antibody produced in rabbit, affinity isolated antibody
SAB4504146
Anti-phospho-Chk1 (pSer286) antibody produced in rabbit, affinity isolated antibody
SAB4504025
Anti-phospho-Chk1 (pSer296) antibody produced in rabbit, affinity isolated antibody
SAB4504147
Anti-phospho-Chk1 (pSer301) antibody produced in rabbit, affinity isolated antibody
SAB4504364
Anti-phospho-Chk1 (pSer317) antibody produced in rabbit, affinity isolated antibody
SAB4504365
Anti-phospho-Chk1 (pSer345) antibody produced in rabbit, affinity isolated antibody
SAB4504366
Anti-phospho-Chk2 (pSer516) antibody produced in rabbit, affinity isolated antibody
SAB4504089
Anti-phospho-Chk2 (pThr383) antibody produced in rabbit, affinity isolated antibody
SAB4503860
Anti-phospho-Chk2 (pThr387) antibody produced in rabbit, affinity isolated antibody
SAB4504367
Anti-phospho-Chk2 (pThr68) antibody produced in rabbit, affinity isolated antibody
SAB1306180 ANTI-PHOSPHO-H3(S10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306179 ANTI-PHOSPHO-H3(S10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305681 ANTI-PHOSPHO-H3(S28) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA008788
Anti-TP53BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution