EMAIL THIS PAGE TO A FRIEND
Thrombosis research
(99m)Tc-ixolaris targets glioblastoma-associated tissue factor: in vitro and pre-clinical applications.
PMID 26070446
Abstract
The clotting initiator protein tissue factor (TF) has recently been described as a potential target that can be exploited to image aggressive tumors. Ixolaris is a specific TF inhibitor that blocks tumor cell procoagulant activity and tumor growth. Herein we evaluated the ability of (99m)Tc-ixolaris to target tumor-derived TF using an orthotopic glioblastoma (GBM) model in mice. The right forebrains of Swiss mice were stereotactically inoculated with U87-MG human GBM cells. Histological and immunohistochemical analyses were performed on the resulting tumors after 35-45 days. The biodistribution of (99m)Tc-ixolaris was evaluated by semi-quantitative whole-body scintigraphy and a quantitative analysis of radioactivity in isolated organs. No (99m)Tc-ixolaris uptake was observed in brain of tumor-free mice, independently of the integrity of brain-blood barrier. In contrast, the presence of TF-expressing brain tumor masses determined a significant (99m)Tc-ixolaris uptake. (99m)Tc-ixolaris recognized TF-expressing GBM cells in vivo. Given the proposed role of TF in tumor progression, (99m)Tc-ixolaris is a promising radiopharmaceutical agent for quantifying cancer-associated TF in aggressive tumors, including GBM.