EMAIL THIS PAGE TO A FRIEND
Inflammation
Analysis of ADRB2 (Arg16Gly) Gene Variant with Susceptibility, Pharmacogenetic Response and Disease Severity in South Indian Asthmatics.
PMID 26071206
Abstract
β2-Adrenergic receptor (β2-AR) plays a crucial role in asthma pathophysiology by regulating, processes of the lung function, and clinical response to bronchodilators. The +46G>A- Gly16Arg polymorphism in the gene encoding β2 adrenergic receptor (ADRB2) has been associated with receptor non-responsiveness after β2-agonist exposure. In the present study, we sought to evaluate the possible association of Gly16Arg polymorphism with asthma susceptibility, pharmacogenetic response to Salbutamol, and varying degrees of disease severity. Three hundred ninety-eight clinically diagnosed patients and 456 healthy controls were enrolled for the study. Patients were classified into severity classes according to Global Initiative for Asthma guidelines. To assess bronchodilator response, spirometry was performed before and 15 min after Salbutamol (200 μg) delivery. Responders to Salbutamol were categorized if percentage reversibility was greater than or equal to 12% in them, while those showing reversibility less than 12% were classified as non-responders. Genotyping was carried out by ARMS-PCR technique. Statistical methods were applied to test for the significance of the results. In the present study, there was lack of significant association of polymorphism with disease susceptibility as well as with bronchodilator response. The polymorphism was not associated with mild and moderate asthma subtypes; however, there was a notable association with severe asthma subtype. In addition, the polymorphism was associated with severe asthma compared to subtypes of mild and moderate asthma combined. In a South Indian population, the ADRB2 Arg/Gly may not form a susceptible variant to develop asthma nor can be a standard predictive marker to bronchodilator response; nevertheless, the patterns in asthma severity can be predicted by analyzing this variant.