EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and evaluation of novel 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives as potent and orally efficacious immunomodulators targeting JAK3.
PMID 26071372
Abstract
Janus kinases (JAKs) regulate various inflammatory and immune responses and are targets for the treatment of inflammatory and immune diseases. As a novel class of immunomodulators targeting JAK3, 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives are promising candidates for treating such diseases. In chemical modification of lead compound 2, the substitution of a cycloalkyl ring for an N-cyanopyridylpiperidine in C4-position was effective for increasing JAK3 inhibitory activity. In addition, modulation of physical properties such as molecular lipophilicity and basicity was important for reducing human ether-a-go-go-related gene (hERG) inhibitory activity. Our optimization study gave compound 31, which exhibited potent JAK3 inhibitory activity as well as weak hERG inhibitory activity. In cellular assay, 31 exhibited potent immunomodulating effect on IL-2-stimulated T cell proliferation. In a pharmacokinetic study, good metabolic stability and oral bioavailability of 31 were achieved in rats, dogs, and monkeys. Further, 31 prolonged graft survival in an in vivo rat heterotopic cardiac transplant model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
297879
Dodecane, anhydrous, ≥99%
C12H26