EMAIL THIS PAGE TO A FRIEND
Scientific reports
Morphine compromises bronchial epithelial TLR2/IL17R signaling crosstalk, necessary for lung IL17 homeostasis.
PMID 26072707
Abstract
Opportunistic lung infection and inflammation is a hallmark of chronic recreational/clinical use of morphine. We show that early induction of IL17 from the bronchial epithelium, following pathogenic encounter is a protective response, which contributes to pathogenic clearance and currently attributed to TLR2 activation in immune cells. Concurrent activation of TLR2 and IL17R in bronchial epithelium results in the sequestration of MyD88 (TLR2 adapter) by Act1/CIKS (IL17R adapter), thereby turning off TLR2 signaling to restore homeostasis. Morphine inhibits the early IL17 release and interaction between Act1 and MyD88, leading to decreased pathogenic clearance and sustained inflammation. Hence, we propose that therapeutically targeting either TLR2 or IL17 in bronchial epithelia, in the context of morphine, can restore inflammatory homeostasis.