EMAIL THIS PAGE TO A FRIEND
Optics express
Ultra-large field-of-view two-photon microscopy.
PMID 26072755
Abstract
We present a two-photon microscope that images the full extent of murine cortex with an objective-limited spatial resolution across an 8 mm by 10 mm field. The lateral resolution is approximately 1 µm and the maximum scan speed is 5 mm/ms. The scan pathway employs large diameter compound lenses to minimize aberrations and performs near theoretical limits. We demonstrate the special utility of the microscope by recording resting-state vasomotion across both hemispheres of the murine brain through a transcranial window and by imaging histological sections without the need to stitch.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9793
Agarose, High EEO