EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
TL1A as a Potential Local Inducer of IL17A Expression in Colon Mucosa of Inflammatory Bowel Disease Patients.
PMID 26072972
Abstract
Interaction between TL1A and death receptor 3 (DR3) co-stimulates T cells, induces production of several pro-inflammatory cytokines and has been linked to pathogenesis of inflammatory bowel disease (IBD). This study aimed to establish a link between expression of TL1A and selected TL1A-induced pro-inflammatory cytokines involved in IBD pathogenesis (IL-4, IL-13, IL-17A and IFN-γ) and to investigate a connection between serum concentration of TL1A in patients with IBD and activation of peripheral blood T cells. Elevated levels of IL-4 (2.91-fold) and IL-13 (4.05-fold) mRNA were detected in the inflamed colon mucosa of patients with ulcerative colitis (UC), IFN-γ mRNA was upregulated (3.23-fold) in the inflamed colon mucosa of patients with Crohn's disease (CD), whereas upregulation of IL-17A and TL1A mRNA was present in the inflamed colon mucosa of patients with both CD and UC (IL-17A: 4.48-fold and 2.74-fold, TL1A: 3.19-fold and 3.22-fold, respectively) vs. control subjects. We did not detect any changes in DR3 mRNA expression in the investigated groups of patients. TL1A mRNA level in colon mucosa of patients with IBD correlated only with the level of IL-17A mRNA but no other investigated cytokines. In colon mucosa, expression of TL1A and DR3 was localized to enterocytes and lamina propria mononuclear cells. We did not find any correlation between serum concentrations of TL1A and IL-17A or changes of CD4(+) or CD8(+) lymphocytes phenotype in patients with IBD. Therefore, our data indicate that TL1A may contribute to pathogenesis of IBD via local but not systemic induction of IL-17A but not IL-4, IL-13 or IFN-γ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500419
Anti-DR3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
D3563
Anti-DR3, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2501411
Anti-GNAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB1400841
Anti-LIPH antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA049079
Anti-LIPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107647
Anti-LIPH antibody produced in rabbit, affinity isolated antibody
HPA021656
Anti-SLC10A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105728
Anti-SLC10A3 antibody produced in rabbit, affinity isolated antibody
SAB3500461
Anti-TL1A antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2102491
Anti-TNFRSF25 antibody produced in rabbit, affinity isolated antibody
SAB1410047
Anti-TNFSF15 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA012948 Anti-TNFSF15 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411713
Anti-TNFSF15 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0008718M6
Monoclonal Anti-TNFRSF25 antibody produced in mouse, clone 4E3, purified immunoglobulin, buffered aqueous solution
SAB1404615
Monoclonal Anti-TNFRSF25 antibody produced in mouse, clone 1H2, purified immunoglobulin, buffered aqueous solution