EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Stability studies of cefoselis sulfate in the solid state.
PMID 26073113
Abstract
The process of degradation was studied by using an HPLC-DAD method. Two degradation products were identified with a hybrid ESI-Q-TOF mass spectrometer. The influence of temperature and relative air humidity (RH) on the stability of cefoselis sulfate was investigated. In the solid state at increased RH the degradation of cefoselis sulfate was an autocatalytic reaction of the first order with respect to substrate concentration while in dry air was first-order reaction depending on the substrate concentration. The kinetic and thermodynamic parameters of degradation were calculated.