EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Solvent Properties of Water in Aqueous Solutions of Elastin-Like Polypeptide.
PMID 26075870
Abstract
The phase-transition temperatures of an elastin-like polypeptide (ELP) with the (GVGVP)40 sequence and solvent dipolarity/polarizability, hydrogen-bond donor acidity, and hydrogen-bond acceptor basicity in its aqueous solutions were quantified in the absence and presence of different salts (Na2SO4, NaCl, NaClO4, and NaSCN) and various osmolytes (sucrose, sorbitol, trehalose, and trimethylamine N-oxide (TMAO)). All osmolytes decreased the ELP phase-transition temperature, whereas NaCl and Na2SO4 decreased, and NaSCN and NaClO4 increased it. The determined phase-transition temperatures may be described as a linear combination of the solvent's dipolarity/polarizability and hydrogen-bond donor acidity. The linear relationship established for the phase-transition temperature in the presence of salts differs quantitatively from that in the presence of osmolytes, in agreement with different (direct and indirect) mechanisms of the influence of salts and osmolytes on the ELP phase-transition temperature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103543
4-Nitroanisole, 97%
C7H7NO3