PloS one
Maintenance of Mouse Nephron Progenitor Cells in Aggregates with Gamma-Secretase Inhibitor.
PMID 26075891
Abstract
Knowledge on how to maintain and expand nephron progenitor cells (NPC) in vitro is important to provide a potentially valuable source for kidney replacement therapies. In our present study, we examined the possibility of optimizing NPC maintenance in the "re-aggregate" system. We found that Six2-expressing (Six2(+))-NPC could be maintained in aggregates reconstituted with dispersed cells from E12.5 mouse embryonic kidneys for at least up to 21 days in culture. The maintenance of Six2(+)-NPC required the presence of ureteric bud cells. The number of Six2(+)-NPC increased by more than 20-fold at day 21, but plateaued after day 14. In an attempt to further sustain NPC proliferation by passage subculture, we found that the new (P1) aggregates reconstituted from the original (P0) aggregates failed to maintain NPC. However, based on the similarity between P1 aggregates and aggregates derived from E15.5 embryonic kidneys, we suspected that the differentiated NPC in P1 aggregates may interfere with NPC maintenance. In support of this notion, we found that preventing NPC differentiation by DAPT, a γ-secretase inhibitor that inhibits Notch signaling pathway, was effective to maintain and expand Six2(+)-NPC in P1 aggregates by up to 65-fold. The Six2(+)-NPC in P1 aggregates retained their potential to epithelialize upon exposure to Wnt signal. In conclusion, we demonstrated in our present study that the "re-aggregation" system can be useful for in vitro maintenance of NPC when combined with γ-secretase inhibitor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
AV32404
Anti-LEF1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32403
Anti-LEF1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500590
Anti-LEF1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002087
Anti-LEF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101332
Anti-LEF1 antibody produced in rabbit, affinity isolated antibody
AV44730
Anti-PODXL (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV44729
Anti-PODXL (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2500809
Anti-PODXL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002110
Anti-PODXL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA045507
Anti-PODXL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA052782
Anti-SIX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103006
Anti-SIX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
B1686
BIO, ≥98% (HPLC)
C16H10BrN3O2
C7855
Monoclonal Anti-Cdh1 antibody produced in mouse, ~2 mg/mL, clone DCS-266, purified immunoglobulin, buffered aqueous solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
C2562
Monoclonal Anti-Cytokeratin, pan (Mixture) antibody produced in mouse, clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2, ascites fluid
L3276 Monoclonal Anti-LEF-1 antibody produced in mouse, ~1 mg/mL, clone 1C3, purified immunoglobulin
WH0051176M3
Monoclonal Anti-LEF1 antibody produced in mouse, clone 5A3, purified immunoglobulin, buffered aqueous solution
SAB1405084
Monoclonal Anti-LEF1 antibody produced in mouse, clone 3H5, purified immunoglobulin, buffered aqueous solution