EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
Yif1B Is Involved in the Anterograde Traffic Pathway and the Golgi Architecture.
PMID 26077767
Abstract
Yif1B is an intracellular membrane-bound protein belonging to the Yip family, shown previously to control serotonin 5-HT1A receptor targeting to dendrites. Because some Yip proteins are involved in the intracellular traffic between the ER and the Golgi, here we investigated the precise localization of Yif1B in HeLa cells. We found that Yif1B is not resident into the Golgi, but rather belongs to the IC compartment. After analyzing the role of Yif1B in protein transport, we showed that the traffic of the VSVG protein marker was accelerated in Yif1B depleted HeLa cells, as well as in hippocampal neurons from Yif1B KO mice. Conversely, Yif1B depletion in HeLa cells did not change the retrograde traffic of ShTx. Interestingly, in long term depletion of Yif1B as in Yif1B KO mice, we observed a disorganized Golgi architecture in CA1 pyramidal hippocampal neurons, which was confirmed by electron microscopy. However, because short term depletion of Yif1B did not change Golgi structure, it is likely that the implication of Yif1B in anterograde traffic does not rely on its role in structural organization of the Golgi apparatus, but rather on its shuttling between the ER, the IC and the Golgi compartments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
563919
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
AlCl3
294713
Aluminum chloride, 99.99% trace metals basis
AlCl3
449598
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
AlCl3
SAB2502094
Anti-CHRM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405619
Anti-CHRM2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029795
Anti-CHRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500245
Anti-CHRM2 antibody produced in rabbit, affinity isolated antibody
SAB4501452
Anti-GRP78, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA038845
Anti-HSPA5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA038846
Anti-HSPA5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21589A Anti-LMAN1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA002320
Anti-LMAN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV46614
Anti-LMAN1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101351
Anti-LMAN1 antibody produced in rabbit, affinity isolated antibody
M9693 Anti-Muscarinic Acetylcholine Receptor M2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T9826
Anti-TGN38 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
T7576
Anti-TGN46 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA012609
Anti-TGOLN2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012723
Anti-TGOLN2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103825
Anti-TGOLN2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0001129M1
Monoclonal Anti-CHRM2 antibody produced in mouse, clone 4B5, purified immunoglobulin, buffered aqueous solution
SAB4200585
Monoclonal Anti-ERGIC-53 antibody produced in mouse, ~1.0 mg/mL, clone ERGIC-3, purified immunoglobulin
SAB5300084
Monoclonal Anti-HSPA5 antibody produced in mouse, clone 4E3, ascites fluid
SAB4100203
Monoclonal Anti-LMAN1 antibody produced in mouse, clone LMAN1 -A157, culture supernatant
SAB4100204
Monoclonal Anti-LMAN1 antibody produced in mouse, clone LMAN1 -B157, culture supernatant
SAB4200235
Monoclonal Anti-TGN46 antibody produced in mouse, ~1 mg/mL, clone TGN46-52, purified immunoglobulin
SAB4200355
Monoclonal Anti-TGN46 antibody produced in mouse, ~1.0 mg/mL, clone TGN46-8, purified immunoglobulin
WH0010618M2
Monoclonal Anti-TGOLN2 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution