EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Development of high internal phase emulsion polymeric monoliths for highly efficient enrichment of trace polycyclic aromatic hydrocarbons from large-volume water samples.
PMID 26077972
Abstract
In this work, polymerized high internal phase emulsion (polyHIPE) monoliths were prepared and applied as monolithic adsorbent materials for proconcentration of trace polycyclic aromatic hydrocarbons (PAHs) from large-volume water samples. The monolithic polyHIPE columns were prepared by in situ polymerization of the continuous phase of a high internal phase emulsion (HIPE) containing styrene (STY), divinylbenzene (DVB) and glycidyl methacrylate (GMA) in pipette tips, and the resulting STY/DVB/GMA polyHIPE monoliths exhibited highly interconnected porosity and large surface areas, making them excellent candidates as adsorbents for enrichment of trace aromatic compounds. The prepared STY/DVB/GMA polyHIPE monoliths were applied to the determination of trace PAHs in environmental water samples by combing with high performance liquid chromatography-fluorescence detection (HPLC-FLD). Under the optimized experimental conditions, the polyHIPE monoliths could effectively enrich trace 13 PAHs from 500mL of water samples, the mean recoveries at four spiked levels were ranged from 80.7% to 115.0% with the relative standard deviations (RSDs) lower than 14%, and the detection limits (LODs) were ranged from 4.0 to 228pg/L. In addition, the prepared polyHIPE monolith was stable enough for more than 200 replicate extraction cycles without measurable loss of performance on the enrichment of PAHs, and good column-to-column repeatability was obtained with RSD less than 13%. The proposed method was applied to simultaneous analysis of 13 PAHs in water samples with satisfactory recoveries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10580
Anthracene, suitable for scintillation, ≥99.0% (GC)
C14H10
331481
Anthracene, zone-refined, ≥99%
C14H10
141062
Anthracene, ReagentPlus®, 99%
C14H10
A89200
Anthracene, reagent grade, 97%
C14H10
694959
Anthracene, sublimed grade, ≥99%
C14H10
ERA-010
Anthracene, vial of 250 mg, analytical standard
C14H10
B2209
Benz[a]anthracene, 99%
C18H12
418099
Benzaldehyde, purified by redistillation, ≥99.5%
C7H6O
B1334
Benzaldehyde, ReagentPlus®, ≥99%
C7H6O
W212709
Benzaldehyde, ≥98%, FG, FCC
C7H6O
W212717
Benzaldehyde, natural, ≥98%, FCC, FG
C7H6O
B1760
Benzo[a]pyrene, ≥96% (HPLC)
C20H12
SCB-002
Benzo[b]fluoranthene, vial of 1 g, analytical standard
C20H12
275336
Benzo[b]fluoranthene, 98%
C20H12
392251
Benzo[k]fluoranthene, for fluorescence, ≥99%
C20H12
245186
Chrysene, 98%
C18H12
SCD-003
Dibenz[a,h]anthracene, vial of 1 g, analytical standard
C22H14
423947
Fluoranthene, 98%
C16H10
F807
Fluoranthene, 98%
C16H10
128333
Fluorene, 98%
C13H10
151238
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
C7H10O3
779342
Glycidyl methacrylate, ≥97.0% (GC)
C7H10O3
184500
Naphthalene, suitable for scintillation, ≥99%
C10H8
147141
Naphthalene, 99%
C10H8
P5592
Potassium peroxodisulfate, BioXtra, ≥99.0%
K2O8S2
240869
Styrene, ReagentPlus®, 99.9%
C8H8
S4972
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
C8H8