EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Host Engineering for High Quantum Efficiency Blue and White Fluorescent Organic Light-Emitting Diodes.
PMID 26078193
Abstract
High quantum efficiency in blue and white fluorescence organic light-emitting diodes is achieved by developing a novel device architecture with fluorescent emitters doped in a thermally activated delayed fluorescent emitter as a host material.