EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Activated NKT Cells Can Condition Different Splenic Dendritic Cell Subsets To Respond More Effectively to TLR Engagement and Enhance Cross-Priming.
PMID 26078270
Abstract
The function of dendritic cells (DCs) can be modulated through multiple signals, including recognition of pathogen-associated molecular patterns, as well as signals provided by rapidly activated leukocytes in the local environment, such as innate-like T cells. In this article, we addressed the possibility that the roles of different murine DC subsets in cross-priming CD8(+) T cells can change with the nature and timing of activatory stimuli. We show that CD8α(+) DCs play a critical role in cross-priming CD8(+) T cell responses to circulating proteins that enter the spleen in close temporal association with ligands for TLRs and/or compounds that activate NKT cells. However, if NKT cells are activated first, then CD8α(-) DCs become conditioned to respond more vigorously to TLR ligation, and if triggered directly, these cells can also contribute to priming of CD8(+) T cell responses. In fact, the initial activation of NKT cells can condition multiple DC subsets to respond more effectively to TLR ligation, with plasmacytoid DCs making more IFN-α and both CD8α(+) and CD8α(-) DCs manufacturing more IL-12. These results suggest that different DC subsets can contribute to T cell priming if provided appropriately phased activatory stimuli, an observation that could be factored into the design of more effective vaccines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1602
1-Decanethiol, 96%
C10H22S
705233
1-Decanethiol, 99%
C10H22S