EMAIL THIS PAGE TO A FRIEND
International journal of medical sciences
Comparison of Retroflexed and Forward Views for Colorectal Endoscopic Submucosal Dissection.
PMID 26078705
Abstract
The use of a retroflexed view exposes the entire tumor surface, which is obscured in the forward view, and contributes to complete tumor resection when combined with forward views. However, the efficacy and safety of using the retroflexed view for colorectal endoscopic submucosal dissection (ESD) are poorly understood. In this study, we assessed the efficacy and safety of the retroflexed view in colorectal ESD. From April 2009 to December 2013, 130 colorectal tumors were examined in 128 patients treated with ESD. A total of 119 patients with a mean tumor size of 27.2 mm were enrolled in the study, and these patients were assigned to undergo colorectal ESD with or without a retroflexed view. The use of retroflexion was successful in 84.2% of patients. There were no perforations in the study and no complications related to the use of retroflexed views. The mean procedure time was 103.6 ± 55.8 min in the retroflexed group, as compared with 108.0 ± 66.5 min in the forward view group. The mean procedure time for resecting tumors >40 mm was significantly shorter in the retroflexed group relative to the forward group. Additionally, the mean dissection speed per unit area was significantly faster in the retroflexed group, as compared with the forward group. Retroflexed views can be used to remove lesions >40 mm and shorten procedure times. Retroflexion may also contribute to an improved en bloc resection rate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P127
(+)-Pentazocine, solid
C19H27NO
P134
(−)-Pentazocine, ≥98% (HPLC)
C19H27NO