EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Increased expression of TLR2 in CD4(+) T cells from SLE patients enhances immune reactivity and promotes IL-17 expression through histone modifications.
PMID 26079624
Abstract
The innate immune system has been shown to play an important pathologic role in systemic lupus erythematosus (SLE). TLR2, a PRR, recognizes exogenous PAMPs, and endogenous damage-associated molecular patterns and has been implicated in the initiation and maintenance of the perpetuated inflammatory reactions in autoimmune diseases. Here, we report increased expression of TLR2 in CD4(+) and CD8(+) T cells, CD19(+) B cells, and CD14(+) monocytes from SLE patients. Conventional treatment, such as hydroxychloroquine and corticosteroids, showed no effect on TLR2 expression in CD4(+) T cells from SLE patients. In vitro stimulation of TLR2 in CD4(+) T cells from SLE patients increased CD40L and CD70 expression, as well as secretion of IL-6, IL-17A, IL-17F, and TNF-α, while Foxp3 transcription decreased. This effect was reversed by TLR2 siRNA. Moreover, TLR2 activation upregulated H3K4 tri-methylation and H4 acetylation levels while downregulated H3K9 tri-methylation level in the IL-17A promoter region. In addition, it also increased H4 acetylation levels and decreased H3K9 tri-methylation levels in the IL-17F promoter region. In summary, our findings demonstrate that increased expression of TLR2 contributes to immune reactivity and promotes IL-17A and IL-17F expression through histone modifications in SLE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409834
Anti-CCDC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019049
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019051
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2107667
Anti-CCDC6 antibody produced in rabbit, affinity isolated antibody
SAB1411455
Anti-CCDC6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410334
Anti-EMP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2107598
Anti-HIST2H4 antibody produced in rabbit, affinity isolated antibody
SAB1300570
Anti-HRH4 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2701955
Anti-HRH4 (N-terminal) antibody produced in rabbit
HPA035009
Anti-HRH4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502598
Anti-HRH4 antibody produced in rabbit, affinity isolated antibody
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306778
ANTI-ICOS(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2102105
Anti-SELS antibody produced in rabbit, affinity isolated antibody
SAB1411081
Anti-SELS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407701
Anti-SELS, (N-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010025
Anti-VIMP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU000321 MISSION® esiRNA, esiRNA targeting human THBD (esiRNA1)
EHU079921 MISSION® esiRNA, esiRNA human MCL1 (esiRNA1)
EMU077471 MISSION® esiRNA, esiRNA targeting mouse Thbd (esiRNA1)
EMU064061 MISSION® esiRNA, esiRNA targeting mouse Mcl1 (esiRNA1)
WH0008030M3
Monoclonal Anti-CCDC6 antibody produced in mouse, clone 5D11, purified immunoglobulin, buffered aqueous solution
WH0002014M1
Monoclonal Anti-EMP3 antibody produced in mouse, clone 3D4, purified immunoglobulin, buffered aqueous solution
SAB1403769
Monoclonal Anti-EMP3 antibody produced in mouse, clone 2C4, purified immunoglobulin, buffered aqueous solution
WH0029851M1 Monoclonal Anti-ICOS antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412835
Monoclonal Anti-ICOS antibody produced in mouse, clone 3G4, purified immunoglobulin, 0.5 mg/mL