EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Structure of the Ergothioneine-Biosynthesis Amidohydrolase EgtC.
PMID 26079795
Abstract
The ubiquitous sulfur metabolite ergothioneine is biosynthesized by oxidative attachment of a sulfur atom to the imidazole ring of Nα-trimethylhistidine. Most actinobacteria, including Mycobacterium tuberculosis, use γ-glutamyl cysteine as a sulfur donor. In subsequent steps the carbon scaffold of γ-glutamyl cysteine is removed by the glutamine amidohydrolase EgtC and the β-lyase EgtE. We determined the crystal structure of EgtC from Mycobacterium smegmatis in complex with its physiological substrate. The set of active site residues that define substrate specificity in EgtC are highly conserved, even in homologues that are not involved in ergothioneine production. This conservation is compounded by the phylogenetic distribution of EgtC-like enzymes indicates that their last common ancestor might have emerged for a purpose other than ergothioneine production.