EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Enhanced physical stabilization of fenofibrate nanosuspensions via wet co-milling with a superdisintegrant and an adsorbing polymer.
PMID 26079832
Abstract
Drug nanoparticles in suspensions can form aggregates leading to physical instability, which is traditionally mitigated using soluble polymers and surfactants. The aim of this paper was to explore common superdisintegrants, i.e., sodium starch glycolate (SSG), croscarmellose sodium (CCS), and crospovidone (CP), as novel class of dispersants for enhanced stabilization of fenofibrate (FNB), a model BCS Class II drug, suspensions. FNB was wet-milled with superdisintegrants along with hydroxypropyl methylcellulose (HPMC), a soluble adsorbing polymer, in a stirred media mill. For comparison, FNB was also milled in the presence of HPMC and/or SDS (sodium dodecyl sulfate) without superdisintegrants. Laser diffraction, scanning electron microscopy, viscometry, differential scanning calorimetry, and powder X-ray diffraction were used to characterize the suspensions. The results show that 2% HPMC along with 1% SSG or 1% CCS mitigated the aggregation of FNB nanoparticles significantly similar to the use of either 5% HPMC or 1% HPMC-0.075% SDS, whereas CP was not effective due to its low swelling capacity. CCS/SSG enhanced steric-kinetic stabilization of the FNB suspensions owing to their high swelling capacity, viscosity enhancement, and physical barrier action. Overall, this study provides a mechanistic basis for a novel method of formulating surfactant-free drug nanosuspensions with co-milled superdisintegrants.