EMAIL THIS PAGE TO A FRIEND
Oncotarget
Tyrosine dephosphorylation enhances the therapeutic target activity of epidermal growth factor receptor (EGFR) by disrupting its interaction with estrogen receptor (ER).
PMID 26079946
Abstract
Protein-protein interactions can increase or decrease its therapeutic target activity and the determining factors involved, however, are largely unknown. Here, we report that tyrosine-dephosphorylation of epidermal growth factor receptor (EGFR) increases its therapeutic target activity by disrupting its interaction with estrogen receptor (ER). Protein tyrosine phosphatase H1 (PTPH1) dephosphorylates the tyrosine kinase EGFR, disrupts its interaction with the nuclear receptor ER, and increases breast cancer sensitivity to small molecule tyrosine kinase inhibitors (TKIs). These effects require PTPH1 catalytic activity and its interaction with EGFR, suggesting that the phosphatase may increase the sensitivity by dephosphorylating EGFR leading to its dissociation with ER. Consistent with this notion, a nuclear-localization defective ER has a higher EGFR-binding activity and confers the resistance to TKI-induced growth inhibition. Additional analysis show that PTPH1 stabilizes EGFR, stimulates the membranous EGFR accumulation, and enhances the growth-inhibitory activity of a combination therapy of TKIs with an anti-estrogen. Since EGFR and ER both are substrates for PTPH1 in vitro and in intact cells, these results indicate that an inhibitory EGFR-ER protein complex can be switched off through a competitive enzyme-substrate binding. Our results would have important implications for the treatment of breast cancer with targeted therapeutics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4504579
Anti-phospho-EGFR (pSer1026) antibody produced in rabbit, affinity isolated antibody
SAB4300062
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504382
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504580
Anti-phospho-EGFR (pSer1071) antibody produced in rabbit, affinity isolated antibody
SAB4504578
Anti-phospho-EGFR (pSer695) antibody produced in rabbit, affinity isolated antibody
SAB4300151
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4503759
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4300152
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
SAB4503760
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
SAB4503909
Anti-phospho-EGFR (pTyr1016) antibody produced in rabbit, affinity isolated antibody
SAB4504170
Anti-phospho-EGFR (pTyr1069) antibody produced in rabbit, affinity isolated antibody
SAB4300063
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4504383
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4300209
Anti-phospho-EGFR (pTyr1110) antibody produced in rabbit, affinity isolated antibody
SAB4504384
Anti-phospho-EGFR (pTyr1110) antibody produced in rabbit, affinity isolated antibody
SAB4300177
Anti-phospho-EGFR (pTyr1172) antibody produced in rabbit, affinity isolated antibody
SAB4504385
Anti-phospho-EGFR (pTyr1172) antibody produced in rabbit, affinity isolated antibody
SAB4300285
Anti-phospho-EGFR (pTyr1197) antibody produced in goat, affinity isolated antibody
SAB4300183
Anti-phospho-EGFR (pTyr1197) antibody produced in rabbit, affinity isolated antibody
SAB4504386
Anti-phospho-EGFR (pTyr1197) antibody produced in rabbit, affinity isolated antibody
SAB4300184
Anti-phospho-EGFR (pTyr869) antibody produced in rabbit, affinity isolated antibody
SAB4504387
Anti-phospho-EGFR (pTyr869) antibody produced in rabbit, affinity isolated antibody