EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
DJ-1 upregulates anti-oxidant enzymes and attenuates hypoxia/re-oxygenation-induced oxidative stress by activation of the nuclear factor erythroid 2-like 2 signaling pathway.
PMID 26081287
Abstract
DJ-1 protein, as a multifunctional intracellular protein, has an important role in transcriptional regulation and anti-oxidant stress. A recent study by our group showed that DJ-1 can regulate the expression of certain anti‑oxidant enzymes and attenuate hypoxia/re‑oxygenation (H/R)‑induced oxidative stress in the cardiomyocyte cell line H9c2; however, the detailed molecular mechanisms have remained to be elucidated. Nuclear factor erythroid 2‑like 2 (Nrf2) is an essential transcription factor that regulates the expression of several anti‑oxidant genes via binding to the anti‑oxidant response element (ARE). The present study investigated whether activation of the Nrf2 pathway is responsible for the induction of anti‑oxidative enzymes by DJ‑1 and contributes to the protective functions of DJ‑1 against H/R‑induced oxidative stress in H9c2 cells. The results demonstrated that DJ‑1‑overexpressing H9c2 cells exhibited anti‑oxidant enzymes, including manganese superoxide dismutase, catalase and glutathione peroxidase, to a greater extent and were more resistant to H/R‑induced oxidative stress compared with native cells, whereas DJ‑1 knockdown suppressed the induction of these enzymes and further augmented the oxidative stress injury. Determination of the importance of Nrf2 in DJ‑1‑mediated anti‑oxidant enzymes induction and cytoprotection against oxidative stress induced by H/R showed that overexpression of DJ‑1 promoted the dissociation of Nrf2 from its cytoplasmic inhibitor Keap1, resulting in enhanced levels of nuclear translocation, ARE‑binding and transcriptional activity of Nrf2. Of note, Nrf2 knockdown abolished the DJ‑1‑mediated induction of anti‑oxidant enzymes and cytoprotection against oxidative stress induced by H/R. In conclusion, these findings indicated that activation of the Nrf2 pathway is a critical mechanism by which DJ-1 upregulates anti-oxidative enzymes and attenuates H/R-induced oxidative stress in H9c2 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32950
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
C13H14N2
T8203
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture
SAB4500248
Anti-DJ-1 antibody produced in rabbit, affinity isolated antibody
SAB1306166 ANTI-DJ-1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
P9373 Anti-PARK7 (DJ-1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004190
Anti-PARK7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101719
Anti-PARK7 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4500249
Anti-PARK7 antibody produced in rabbit, affinity isolated antibody
D2385
Diphenylamine, ≥98%
C12H11N
242586
Diphenylamine, ACS reagent, ≥99%
C12H11N
112763
Diphenylamine, ReagentPlus®, 99%
C12H11N
E1510
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
C21H20BrN3
E1385
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
C21H20BrN3
46067
Ethidium bromide solution, for fluorescence, ~1% in H2O
C21H20BrN3
F2555
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid
46047
Pyridaben, PESTANAL®, analytical standard
C19H25ClN2OS