EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Modified quaternary ammonium salts as potential antimalarial agents.
PMID 26081764
Abstract
A series of new quaternary ammonium salts containing a polyconjugated moiety has been synthesized and characterized; their biological activity as potential antimalarial agents was investigated, as well. All compounds were screened against chloroquine resistant W-2 (CQ-R) and chloroquine sensitive, D-10 (CQ-S) strains of Plasmodium falciparum showing IC50 in the submicromolar range and low toxicity against human endothelial cells.