EMAIL THIS PAGE TO A FRIEND
Cancer research
Interleukin-6 Stimulates Defective Angiogenesis.
PMID 26081809
Abstract
The cytokine IL6 has a number of tumor-promoting activities in human and experimental cancers, but its potential as an angiogenic agent has not been fully investigated. Here, we show that IL6 can directly induce vessel sprouting in the ex vivo aortic ring model, as well as endothelial cell proliferation and migration, with similar potency to VEGF. However, IL6-stimulated aortic ring vessel sprouts had defective pericyte coverage compared with VEGF-stimulated vessels. The mechanism of IL6 action on pericytes involved stimulation of the Notch ligand Jagged1 as well as angiopoietin2 (Ang2). When peritoneal xenografts of ovarian cancer were treated with an anti-IL6 antibody, pericyte coverage of vessels was restored. In addition, in human ovarian cancer biopsies, there was an association between levels of IL6 mRNA, Jagged1, and Ang2. Our findings have implications for the use of cancer therapies that target VEGF or IL6 and for understanding abnormal angiogenesis in cancers, chronic inflammatory disease, and stroke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA002951
Anti-CSPG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA042785
Anti-CSPG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005928
Anti-EMCN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700787
Anti-EMCN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502163
Anti-Mucin-14, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2102034
Anti-RNF5 antibody produced in rabbit, affinity isolated antibody
SAB4200208
Anti-RNF5 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2701502
Anti-RNF5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1408906
Anti-VCAN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004726 Anti-VCAN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
L9381 Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), FITC conjugate, lyophilized powder
L5264 Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), TRITC conjugate, lyophilized powder
N8912 Monoclonal Anti-NG2 antibody produced in mouse, clone 132.38, purified immunoglobulin, buffered aqueous solution
R0278 RIPA Buffer