EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
KLF4 mediates the link between TGF-β1-induced gene transcription and H3 acetylation in vascular smooth muscle cells.
PMID 26082460
Abstract
Transcriptional activation by transcription factors is coupled with histone acetylation and chromatin remodeling. However, the relationship between TGF-β1-induced gene transcription by Krüppel-like factor (KLF)-4 and histone acetylation remains unknown. In our study, KLF4 overexpression or knockdown, respectively increased or decreased H3 acetylation and p300 occupancy, which is concentrated in the region containing TGF-β1 control elements (TCEs) of the genes by TGF-β1 regulation during vascular smooth muscle cell (VSMC) differentiation. Coimmunoprecipitation and glutathione S-transferase pull-down assays showed that phosphatase and tensin homolog (PTEN) formed a complex with KLF4 to inhibit the phosphorylation of the latter in basal conditions. After TGF-β1 signaling activation, PTEN was phosphorylated by p38 MAPK or PI3K/Akt signaling, phosphorylated PTEN lost its ability to dephosphorylate KLF4, and the cofactors interacting with KLF4 switched from PTEN to p300. Then, KLF4-p300 complexes were recruited to KLF4-binding sites of the gene promoter of VSMCs, to acetylate histone H3 and activate transcription. In addition, phosphorylated KLF4 enhanced p300 histone acetyltransferase (HAT) activity via the p38 MAPK pathway, which may be responsible for H3 acetylation. Taken together, the results of our study reveal a novel mechanism whereby KLF4 mediates the link between TGF-β1-induced gene transcription activation and H3 acetylation during VSMC differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003128
Anti-EP300 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004112
Anti-EP300 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV38430
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV35721
Anti-KLF4 antibody produced in rabbit, IgG fraction of antiserum
HPA002926
Anti-KLF4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1300678
Anti-KLF4 antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB2105308
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody
SAB2701975
Anti-KLF4 antibody produced in rabbit
SAB2106435
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody
SAB4503485
Anti-p300, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4504520
Anti-phospho-PTEN (pSer370) antibody produced in rabbit, affinity isolated antibody
SAB4300044
Anti-phospho-PTEN (pSer380/pThr382/pThr383) antibody produced in rabbit, affinity isolated antibody
SAB4504521
Anti-phospho-PTEN (pSer380/Thr382/383) antibody produced in rabbit, affinity isolated antibody
SAB4503980
Anti-phospho-PTEN (pSer385) antibody produced in rabbit, affinity isolated antibody
SAB4504207
Anti-phospho-Smad2 (pSer250) antibody produced in rabbit, affinity isolated antibody
SAB4300252
Anti-phospho-SMAD2 (pThr220) antibody produced in rabbit, affinity isolated antibody
SAB4300563
Anti-SMAD2 (Ab-220) antibody produced in rabbit, affinity isolated antibody
SAB4300562
Anti-SMAD2 (Ab-467) antibody produced in rabbit, affinity isolated antibody
SAB4200354
Anti-SMAD2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2702053
Anti-SMAD2 antibody produced in rabbit
SAB4501805
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501806
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501807
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501808
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB2103328
Anti-SMAD2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4501804
Anti-SMAD2, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0002033M2
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1400094
Monoclonal Anti-EP300 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB1403776
Monoclonal Anti-EP300 antibody produced in mouse, clone 1D2, ascites fluid
SAB5300069
Monoclonal Anti-KLF4 antibody produced in mouse, clone 1E6, ascites fluid