EMAIL THIS PAGE TO A FRIEND
Circulation research
B-1b Cells Secrete Atheroprotective IgM and Attenuate Atherosclerosis.
PMID 26082558
Abstract
B cells contribute to atherosclerosis through subset-specific mechanisms. Whereas some controversy exists about the role of B-2 cells, B-1a cells are atheroprotective because of secretion of atheroprotective IgM antibodies independent of antigen. B-1b cells, a unique subset of B-1 cells that respond specifically to T-cell-independent antigens, have not been studied within the context of atherosclerosis. To determine whether B-1b cells produce atheroprotective IgM antibodies and function to protect against diet-induced atherosclerosis. We demonstrate that B-1b cells are sufficient to produce IgM antibodies against oxidation-specific epitopes on low-density lipoprotein both in vitro and in vivo. In addition, we demonstrate that B-1b cells provide atheroprotection after adoptive transfer into B- and T-cell deficient (Rag1(-/-)Apoe(-/-)) hosts. We implicate inhibitor of differentiation 3 (Id3) in the regulation of B-1b cells as B-cell-specific Id3 knockout mice (Id3(BKO)Apoe(-/-)) have increased numbers of B-1b cells systemically, increased titers of oxidation-specific epitope-reactive IgM antibodies, and significantly reduced diet-induced atherosclerosis when compared with Id3(WT)Apoe(-/-) controls. Finally, we report that the presence of a homozygous single nucleotide polymorphism in ID3 in humans that attenuates Id3 function is associated with an increased percentage of circulating B-1 cells and anti-malondialdehyde-low-density lipoprotein IgM suggesting clinical relevance. These results provide novel evidence that B-1b cells produce atheroprotective oxidation-specific epitope-reactive IgM antibodies and protect against atherosclerosis in mice and suggest that similar mechanisms may occur in humans.