PloS one
First Demonstration of Antigen Induced Cytokine Expression by CD4-1+ Lymphocytes in a Poikilotherm: Studies in Zebrafish (Danio rerio).
PMID 26083432
Abstract
Adaptive immunity in homeotherms depends greatly on CD4+ Th cells which release cytokines in response to specific antigen stimulation. Whilst bony fish and poikilothermic tetrapods possess cells that express TcR and CD4-related genes (that exist in two forms in teleost fish; termed CD4-1 and CD4-2), to date there is no unequivocal demonstration that cells equivalent to Th exist. Thus, in this study we determined whether CD4-1+ lymphocytes can express cytokines typical of Th cells following antigen specific stimulation, using the zebrafish (Danio rerio). Initially, we analyzed the CD4 locus in zebrafish and found three CD4 homologues, a CD4-1 molecule and two CD4-2 molecules. The zfCD4-1 and zfCD4-2 transcripts were detected in immune organs and were most highly expressed in lymphocytes. A polyclonal antibody to zfCD4-1 was developed and used with an antibody to ZAP70 and revealed double positive cells by immunohistochemistry, and in the Mycobacterium marinum disease model CD4-1+ cells were apparent surrounding the granulomas typical of the infection. Next a prime-boost experiment, using human gamma globulin as antigen, was performed and revealed for the first time in fish that zfCD4-1+ lymphocytes increase the expression of cytokines and master transcription factors relevant to Th1/Th2-type responses as a consequence of boosting with specific antigen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36001
Anti-CREBZF antibody produced in rabbit, IgG fraction of antiserum
SAB2100482
Anti-CREBZF antibody produced in rabbit, affinity isolated antibody
SAB2105692
Anti-CREBZF antibody produced in rabbit, affinity isolated antibody
HPA021526
Anti-CREBZF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502556
Anti-CREBZF, C-Terminal antibody produced in rabbit, affinity isolated antibody
F7512 Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB4503106
Anti-ZAP-70 antibody produced in rabbit, affinity isolated antibody
SAB4503107
Anti-ZAP-70 antibody produced in rabbit, affinity isolated antibody
SAB4503108
Anti-ZAP-70 antibody produced in rabbit, affinity isolated antibody
SAB4503109
Anti-ZAP-70, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300437
Anti-ZAP70 (Ab-319) antibody produced in rabbit, affinity isolated antibody
SAB4300438
Anti-ZAP70 (Ab-493) antibody produced in rabbit, affinity isolated antibody
SAB2502042
Anti-ZAP70 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA003134
Anti-ZAP70 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107611
Anti-ZAP70 antibody produced in rabbit, affinity isolated antibody
EHU091301 MISSION® esiRNA, esiRNA human ZAP70 (esiRNA1)
Z0627
Monoclonal Anti-ZAP-70 antibody produced in mouse, ~1.5 mg/mL, clone 1E7.2, purified immunoglobulin, buffered aqueous solution
SAB4700528 Monoclonal Anti-ZAP70 , (C-terminal) antibody produced in mouse, clone ZAP-03, purified immunoglobulin, buffered aqueous solution
WH0007535M1
Monoclonal Anti-ZAP70 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
SRP0338
ZAP70 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)