PloS one
Clinical Impact of Rapid Reduction of Low-Density Lipoprotein Cholesterol Level on Long-Term Outcome of Acute Myocardial Infarction in the Statin Era: Subanalysis of the ALPS-AMI Study.
PMID 26083546
Abstract
The optimal period to achieve target percent reduction of low-density lipoprotein cholesterol (LDL-C) level for secondary prevention of acute myocardial infarction (AMI) is not well established. The Assessment of Lipophilic vs. Hydrophilic Statin Therapy in AMI (ALPS-AMI) study enrolled 508 patients (mean age, 66.0± 11.6 years; 80.6% male) who were hospitalized for AMI and underwent percutaneous coronary intervention (PCI). Of these patients, 81 were excluded because of the absence of LDL-C measurements at 4 weeks after randomization. In the remaining 427 patients, the target LDL-C level reduction of ≥30% was achieved and not reached within 4 weeks after randomization in 204 cases (early reduction group) and 223 cases (late reduction group). The groups were formed prospectively and analyzed with regard to the composite end point (major adverse cardiovascular event [MACE]: all-cause death, myocardial infarction, and stroke) and clinical outcomes. MACE were significantly more frequent in the late reduction group compared to the early reduction group (9.4% vs. 3.4%, P = 0.013). The incidence of cardiac deaths was also significantly higher in the late reduction group (3.1% vs. 0.5%, P = 0.044). On age-adjusted Cox proportional hazards analysis in statin-naïve patients, percent reduction of LDL-C level during the initial 4 weeks (HR, 0.98; 95% CI: 0.97-0.99, P = 0.042) and baseline LDL-C level (HR, 0.98; 95% CI: 0.97-0.99, P = 0.033) predicted adverse events. Rapid reduction of LDL-C level is strongly associated with favorable outcome in patients with AMI.