EMAIL THIS PAGE TO A FRIEND
Arquivos de neuro-psiquiatria
Effect of volume replacement during combined experimental hemorrhagic shock and traumatic brain injury in prostanoids, brain pathology and pupil status.
PMID 26083885
Abstract
Traumatic brain injury (TBI) is the main cause of trauma-related deaths. Systemic hypotension and intracranial hypertension causes cerebral ischemia by altering metabolism of prostanoids. We describe prostanoid, pupilar and pathological response during resuscitation with hypertonic saline solution (HSS) in TBI. Method Fifteen dogs were randomized in three groups according to resuscitation after TBI (control group; lactated Ringer's (LR) group and HSS group), with measurement of thromboxane, prostaglandin, macroscopic and microscopic pathological evaluation and pupil evaluation.Result Concentration of prostaglandin is greater in the cerebral venous blood than in plasma and the opposite happens with concentration of thromboxane. Pathology revealed edema in groups with the exception of group treated with HSS.Discussion and conclusion There is a balance between the concentrations of prostaglandin and thromboxane. HSS prevented the formation of cerebral edema macroscopically detectable. Pupillary reversal occurred earlier in HSS group than in LR group.