EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Remazol-Catalyzed Hydroperoxyarylation of Styrenes.
PMID 26084245
Abstract
A mild photocatalytic hydroperoxyarylation of styrenes has been developed, in which a novel photocatalyst, remazol brilliant blue R (RBBR), is employed at low catalytic loading (1 mol %). The operationally easy procedure uses air as the dioxygen source. Simple mono-substituted styrenes react with aryl hydrazines in moderate-to-good yields. RBBR is proposed to act as a photosensitizer for the generation of singlet oxygen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R8001
Remazol Brilliant Blue R, anthraquinone dye
C22H16N2Na2O11S3