EMAIL THIS PAGE TO A FRIEND
Science translational medicine
Transglutaminase 4 as a prostate autoantigen in male subfertility.
PMID 26084804
Abstract
Autoimmune polyendocrine syndrome type 1 (APS1), a monogenic disorder caused by AIRE gene mutations, features multiple autoimmune disease components. Infertility is common in both males and females with APS1. Although female infertility can be explained by autoimmune ovarian failure, the mechanisms underlying male infertility have remained poorly understood. We performed a proteome-wide autoantibody screen in APS1 patient sera to assess the autoimmune response against the male reproductive organs. By screening human protein arrays with male and female patient sera and by selecting for gender-imbalanced autoantibody signals, we identified transglutaminase 4 (TGM4) as a male-specific autoantigen. Notably, TGM4 is a prostatic secretory molecule with critical role in male reproduction. TGM4 autoantibodies were detected in most of the adult male APS1 patients but were absent in all the young males. Consecutive serum samples further revealed that TGM4 autoantibodies first presented during pubertal age and subsequent to prostate maturation. We assessed the animal model for APS1, the Aire-deficient mouse, and found spontaneous development of TGM4 autoantibodies specifically in males. Aire-deficient mice failed to present TGM4 in the thymus, consistent with a defect in central tolerance for TGM4. In the mouse, we further link TGM4 immunity with a destructive prostatitis and compromised secretion of TGM4. Collectively, our findings in APS1 patients and Aire-deficient mice reveal prostate autoimmunity as a major manifestation of APS1 with potential role in male subfertility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701274
Anti-Human IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701284 Anti-Human IgG (Fc specific), highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701289 Anti-Human IgG (γ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
I9885 Anti-Human IgG (γ-chain specific), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701314 Anti-Human IgG (H+L), F(ab) fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701247 Anti-Human IgG F(ab′)2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701246 Anti-Human IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501703
Anti-TGM4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406527
Anti-TGM4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA032072
Anti-TGM4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104763
Anti-TGM4, (N-terminal) antibody produced in rabbit, affinity isolated antibody