EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Effect of Nitric Oxide Synthesis Blockade on the Morphology of Langerhans Islets in August and Wistar Rats with Acute Alloxan Diabetes.
PMID 26085363
Abstract
Alloxan diabetes was modeled in August rats with high activity of the NO system and in Wistar rats, and the effects of NO system blockade (by a course treatment with L-NNA) on Langerhans islet β cells were studied in 15 days. The toxic effects of diabetes on the rat β cells and islets were similar: the content of active β cells in the islets decreased to 15-20%, the number of islets to 24-29% of control. A course of L-NNA reduced the β cell and islet death, in August cells greater than in Wistar: the number of islets in August rats was restored to 81%, in Wistar rats to 60% of initial level; the activity of β cells remained at the control level in the former and 2-fold lower than in the control in the latter. It seems that a less pronounced protective effect of L-NNA in Wistar rats was explained by excessive reduction of NO level essential for β cell regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5501
Nω-Nitro-L-arginine, ≥98% (TLC)
C6H13N5O4