EMAIL THIS PAGE TO A FRIEND
Acta chimica Slovenica
Synthesis and Structural Study of Some Pyrimidinium Hexafluoridosilicates.
PMID 26085410
Abstract
The synthesis and spectroscopic characterization of some pyrimidinium hexafluoridosilicate salts with the formula (RH)2[SiF]6 (where R = 2-aminopyrimidine (1), cytosine (2) and 5-cyanocytosine (3)) is obtained in the reaction of SiO2 with hydrofluoric acid solution and subsequent addition of 2-aminopyrimidine, cytosine or 4-amino-2-chloropyrimidine-5-carbonitrile. The crystal structures of 1 and 3 have been determined by single-crystal X‒ray diffraction analysis. The combination of N‒H···F hydrogen bonds along with π‒π interactions between aromatic rings resulted in supramolecular architecture of cations and anions in the solid state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78608
2-Aminopyrimidine, 97%
C4H5N3
597600
4-Amino-2-chloropyrimidine-5-carbonitrile, 97%
C5H3ClN4