EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Lack of GCN5 remarkably enhances the resistance against prolonged endoplasmic reticulum stress-induced apoptosis through up-regulation of Bcl-2 gene expression.
PMID 26086109
Abstract
The endoplasmic reticulum (ER), a complex membrane structure, has important roles in all eukaryotic cells. Catastrophe of its functions would lead to ER stress that causes various diseases such as cancer, neurodegenerative diseases, diabetes and so on. Prolonged ER stress could trigger apoptosis via activation of various signal transduction pathways. To investigate physiological roles of histone acetyltransferase GCN5 in regulation of ER stress, we analyzed responses of homozygous GCN5-deficient DT40 mutants, ΔGCN5, against ER stress. GCN5-deficiency in DT40 caused drastic resistance against apoptosis induced by pharmacological ER stress agents (thapsigargin and tunicamycin). Pharmaceutical analysis using specific Bcl-2 inhibitors showed that the drastic resistance against prolonged ER stress-induced apoptosis is, in part, due to up-regulation of Bcl-2 gene expression in ΔGCN5. These data revealed that GCN5 is involved in regulation of prolonged ER stress-induced apoptosis through controlling Bcl-2 gene expression.